Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198923.2 | NP_001185852.2 | 5 - 6 | 948 | 75 | |
GenBank mRNA | AK393630.1 | - | 5 - 6 | 75 | ||
AK395655.1 | - | 5 - 6 | 75 | |||
EU715290.1 | - | 5 - 6 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |