Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204935.1 | NP_001191864.1 | 3 - 4 | 243 | 92 | |
XM_005664980.3 | XP_005665037.1 | 328 | 92 | |||
GenBank mRNA | CK458744.1 | - | 3 - 4 | 243 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |