Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097465.1 | NP_001090934.1 | 4 - 5 | 237 | 120 | |
GenBank mRNA | AK344769.1 | - | 4 - 5 | 272 | 120 | |
AY609707.2 | - | 341 | 120 | |||
DQ629140.1 | - | 4 - 5 | 237 | 120 | ||
DQ629141.1 | - | 4 - 5 | 237 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |