Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161644.2 | NP_001155116.2 | 4 - 5 | 820 | 100 | |
XM_021068342.1 | XP_020924001.1 | 763 | 100 | |||
XM_021068343.1 | XP_020924002.1 | 745 | 100 | |||
GenBank mRNA | FJ873105.1 | - | 4 - 5 | 820 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |