Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244980.1 | NP_001231909.1 | 4 - 5 | 396 | 91 | |
XM_005674019.3 | XP_005674076.1 | 499 | 91 | |||
GenBank mRNA | AK230843.1 | - | 4 - 5 | 396 | 91 | |
AK392033.1 | - | 4 - 5 | 409 | 91 | ||
AK399546.1 | - | 4 - 5 | 419 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |