Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244987.1 | NP_001231916.1 | 6 - 7 | 790 | 101 | |
XM_005655748.3 | XP_005655805.1 | 827 | 101 | |||
GenBank mRNA | AK234481.1 | - | 6 - 7 | 790 | 101 | |
AK346168.1 | - | 6 - 7 | 724 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |