Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245976.1 | NP_001232905.1 | 3 - 4 | 435 | 76 | |
XM_021068454.1 | XP_020924113.1 | 499 | 76 | |||
XM_021068455.1 | XP_020924114.1 | 776 | 76 | |||
XM_021068456.1 | XP_020924115.1 | 569 | 76 | |||
XM_021068457.1 | XP_020924116.1 | 532 | 76 | |||
XM_021068458.1 | XP_020924117.1 | 872 | 76 | |||
XM_021068459.1 | XP_020924118.1 | 462 | 76 | |||
GenBank mRNA | AK393600.1 | - | 3 - 4 | 430 | 76 | |
JF271685.1 | - | 3 - 4 | 435 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |