Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206438.1 | NP_001193367.1 | 2 - 3 | 204 | 65 | |
GenBank mRNA | BX920898.2 | - | 2 - 3 | 191 | 65 | |
BX924569.2 | - | 2 - 3 | 207 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |