Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243255.1 | NP_001230184.1 | 4 - 5 | 714 | 103 | |
XM_021077227.1 | XP_020932886.1 | 667 | 103 | |||
GenBank mRNA | CV874307.1 | - | 4 - 5 | 518 | 103 | |
ES446305.1 | - | 4 - 5 | 542 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |