Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243395.1 | NP_001230324.1 | 3 - 4 | 301 | 76 | |
GenBank mRNA | AK235887.1 | - | 3 - 4 | 331 | 76 | |
FD627892.1 | - | 3 - 4 | 301 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |