Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243412.1 | NP_001230341.1 | 10 - 11 | 1395 | 66 | |
NM_001243413.1 | NP_001230342.1 | 9 - 10 | 1252 | 66 | ||
XM_005655700.3 | XP_005655757.1 | 2035 | 66 | |||
XM_013988430.2 | XP_013843884.1 | 2120 | 66 | |||
XM_013988431.2 | XP_013843885.1 | 1294 | 66 | |||
XM_021090919.1 | XP_020946578.1 | 2041 | 66 | |||
XM_021090921.1 | XP_020946580.1 | 1284 | 66 | |||
XM_021090922.1 | XP_020946581.1 | 2120 | 66 | |||
XM_021090923.1 | XP_020946582.1 | 1109 | 66 | |||
XM_021090924.1 | XP_020946583.1 | 1846 | 66 | |||
GenBank mRNA | AK343836.1 | - | 10 - 11 | 66 | ||
AK400126.1 | - | 10 - 11 | 1378 | 66 | ||
BP443627.1 | - | 6 - 7 | 780 | 66 | ||
EW379696.2 | - | 2 - 3 | 176 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |