Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243441.1 | NP_001230370.1 | 4 - 5 | 642 | 96 | |
GenBank mRNA | AK231739.1 | - | 4 - 5 | 642 | 96 | |
AK232111.1 | - | 4 - 5 | 632 | 96 | ||
AK348629.1 | - | 4 - 5 | 591 | 96 | ||
AK350256.1 | - | 4 - 5 | 610 | 96 | ||
AK352279.1 | - | 4 - 5 | 618 | 96 | ||
AK397511.1 | - | 4 - 5 | 637 | 96 | ||
AY609894.1 | - | 4 - 5 | 620 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |