Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243470.1 | NP_001230399.1 | 11 - 12 | 1181 | 91 | |
GenBank mRNA | AK235362.1 | - | 10 - 11 | 1135 | 91 | |
AK346494.1 | - | 11 - 12 | 1181 | 91 | ||
AK394808.1 | - | 10 - 11 | 1204 | 91 | ||
AY610211.1 | - | 1048 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |