Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243501.1 | NP_001230430.1 | 2 - 3 | 1145 | 72 | |
GenBank mRNA | AK343472.1 | - | 2 - 3 | 1176 | 72 | |
CJ007023.1 | - | 1 - 2 | 279 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |