Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243581.1 | NP_001230510.1 | 2 - 3 | 316 | 92 | |
GenBank mRNA | AK232873.1 | - | 2 - 3 | 322 | 92 | |
AK345202.1 | - | 2 - 3 | 316 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |