Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSNAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243607.2 | NP_001230536.1 | 2 - 3 | 230 | 88 | |
GenBank mRNA | AK344359.1 | - | 2 - 3 | 217 | 88 | |
EW144088.2 | - | 2 - 3 | 271 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |