Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243622.1 | NP_001230551.1 | 1 - 2 | 236 | 82 | |
XM_021100397.1 | XP_020956056.1 | 384 | 82 | |||
XM_021100398.1 | XP_020956057.1 | 392 | 82 | |||
GenBank mRNA | AK232809.1 | - | 1 - 2 | 236 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |