Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNTTIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243674.1 | NP_001230603.1 | 6 - 7 | 2236 | 67 | |
GenBank mRNA | AK344464.1 | - | 6 - 7 | 2250 | 67 | |
CF368239.1 | - | 1 - 2 | 298 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |