Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243691.1 | NP_001230620.1 | 8 - 9 | 1477 | 97 | |
GenBank mRNA | AK349998.1 | - | 8 - 9 | 1477 | 97 | |
AK392015.1 | - | 7 - 8 | 1392 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |