Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243709.1 | NP_001230638.1 | 1 - 2 | 206 | 71 | |
GenBank mRNA | AK351364.1 | - | 1 - 2 | 206 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |