Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RESF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243821.1 | NP_001230750.1 | 4 - 5 | 5236 | 88 | |
XM_005664006.3 | XP_005664063.1 | 5274 | 88 | |||
GenBank mRNA | CN155723.1 | - | 1 - 2 | 449 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Sus scrofa Probe spans exons |