Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243905.1 | NP_001230834.1 | 5 - 6 | 517 | 123 | |
XM_021068362.1 | XP_020924021.1 | 184 | 123 | |||
GenBank mRNA | AK232815.1 | - | 5 - 6 | 517 | 123 | |
AY609754.1 | - | 5 - 6 | 516 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Sus scrofa Probe spans exons |