Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243922.1 | NP_001230851.1 | 2 - 3 | 383 | 68 | |
GenBank mRNA | AK239791.1 | - | 2 - 3 | 383 | 68 | |
AK345530.1 | - | 2 - 3 | 380 | 68 | ||
AK345679.1 | - | 2 - 3 | 320 | 68 | ||
AK399713.1 | - | 2 - 3 | 398 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |