Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244142.1 | NP_001231071.1 | 4 - 5 | 266 | 60 | |
NM_001244143.1 | NP_001231072.1 | 4 - 5 | 258 | 60 | ||
GenBank mRNA | AK231770.1 | - | 4 - 5 | 254 | 60 | |
AK345103.1 | - | 4 - 5 | 250 | 60 | ||
AK392529.1 | - | 4 - 5 | 251 | 60 | ||
AK395791.1 | - | 4 - 5 | 250 | 60 | ||
AK398866.1 | - | 4 - 5 | 250 | 60 | ||
AK399411.1 | - | 4 - 5 | 252 | 60 | ||
AK400616.1 | - | 4 - 5 | 250 | 60 | ||
AK400684.1 | - | 4 - 5 | 298 | 60 | ||
EW289129.2 | - | 4 - 5 | 242 | 60 | ||
EW520500.2 | - | 4 - 5 | 298 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |