Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244151.1 | NP_001231080.1 | 3 - 4 | 199 | 176 | |
XM_021098047.1 | XP_020953706.1 | 170 | 176 | |||
GenBank mRNA | AK235262.1 | - | 3 - 4 | 190 | 176 | |
AK345545.1 | - | 1 - 2 | 83 | 176 | ||
AK389202.1 | - | 3 - 4 | 195 | 176 | ||
AK391703.1 | - | 3 - 4 | 173 | 176 | ||
AY609681.1 | - | 3 - 4 | 177 | 176 | ||
FD588173.1 | - | 3 - 4 | 208 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |