Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244161.1 | NP_001231090.1 | 5 - 6 | 578 | 83 | |
GenBank mRNA | AK230948.1 | - | 5 - 6 | 556 | 83 | |
AK231073.1 | - | 5 - 6 | 542 | 83 | ||
AK231135.1 | - | 5 - 6 | 578 | 83 | ||
AK343627.1 | - | 5 - 6 | 542 | 83 | ||
AK343843.1 | - | 5 - 6 | 560 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |