Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SORD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244162.1 | NP_001231091.1 | 8 - 9 | 1067 | 77 | |
XM_021062686.1 | XP_020918345.1 | 929 | 77 | |||
GenBank mRNA | AK236025.1 | - | 8 - 9 | 1027 | 77 | |
AK236181.1 | - | 7 - 8 | 982 | 77 | ||
AK351174.1 | - | 7 - 8 | 949 | 77 | ||
AK389436.1 | - | 8 - 9 | 1067 | 77 | ||
AK392246.1 | - | 8 - 9 | 1055 | 77 | ||
AY610034.1 | - | 8 - 9 | 982 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |