Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244176.1 | NP_001231105.1 | 1 - 2 | 245 | 83 | |
XM_021078525.1 | XP_020934184.1 | 252 | 83 | |||
XM_021078526.1 | XP_020934185.1 | 273 | 83 | |||
XM_021078527.1 | XP_020934186.1 | 252 | 83 | |||
XM_021078528.1 | XP_020934187.1 | 252 | 83 | |||
XM_021078529.1 | XP_020934188.1 | 251 | 83 | |||
XM_021078530.1 | XP_020934189.1 | 273 | 83 | |||
GenBank mRNA | AK231938.1 | - | 1 - 2 | 230 | 83 | |
AK234688.1 | - | 1 - 2 | 182 | 83 | ||
AK348653.1 | - | 1 - 2 | 184 | 83 | ||
AK393133.1 | - | 1 - 2 | 233 | 83 | ||
AK398750.1 | - | 1 - 2 | 259 | 83 | ||
AY610152.1 | - | 1 - 2 | 245 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |