Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244288.1 | NP_001231217.1 | 2 - 3 | 559 | 88 | |
NM_001244289.1 | NP_001231218.1 | 3 - 4 | 607 | 88 | ||
NM_001244290.1 | NP_001231219.1 | 2 - 3 | 506 | 88 | ||
XM_005665296.3 | XP_005665353.1 | 643 | 88 | |||
XM_005665298.3 | XP_005665355.1 | 557 | 88 | |||
GenBank mRNA | AK233501.1 | - | 3 - 4 | 607 | 88 | |
AK234190.1 | - | 2 - 3 | 559 | 88 | ||
AK238776.1 | - | 3 - 4 | 466 | 88 | ||
AK240577.1 | - | 2 - 3 | 506 | 88 | ||
AK395093.1 | - | 2 - 3 | 559 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Inventoried Probe spans exons |