Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244410.1 | NP_001231339.1 | 1 - 2 | 332 | 57 | |
XM_021088538.1 | XP_020944197.1 | 317 | 57 | |||
GenBank mRNA | AK240091.1 | - | 1 - 2 | 332 | 57 | |
AK352158.1 | - | 1 - 2 | 192 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |