Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244436.1 | NP_001231365.1 | 2 - 3 | 342 | 97 | |
XM_005670816.3 | XP_005670873.1 | 339 | 97 | |||
XM_005670820.3 | XP_005670877.1 | 333 | 97 | |||
XM_013983049.2 | XP_013838503.1 | 334 | 97 | |||
GenBank mRNA | AK234715.1 | - | 2 - 3 | 285 | 97 | |
AK240444.1 | - | 2 - 3 | 291 | 97 | ||
AK398652.1 | - | 2 - 3 | 291 | 97 | ||
AK398863.1 | - | 2 - 3 | 282 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |