Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244436.1 | NP_001231365.1 | 3 - 4 | 563 | 52 | |
XM_005670816.3 | XP_005670873.1 | 560 | 52 | |||
XM_005670817.3 | XP_005670874.1 | 516 | 52 | |||
XM_005670818.3 | XP_005670875.1 | 527 | 52 | |||
XM_005670819.3 | XP_005670876.1 | 345 | 52 | |||
XM_013983049.2 | XP_013838503.1 | 555 | 52 | |||
XM_021071885.1 | XP_020927544.1 | 857 | 52 | |||
GenBank mRNA | AK234715.1 | - | 3 - 4 | 506 | 52 | |
AK240444.1 | - | 3 - 4 | 512 | 52 | ||
AK398652.1 | - | 3 - 4 | 512 | 52 | ||
AK398863.1 | - | 3 - 4 | 503 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |