Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244502.1 | NP_001231431.1 | 2 - 3 | 254 | 86 | |
XM_005659420.3 | XP_005659477.2 | 253 | 86 | |||
XM_005659421.3 | XP_005659478.2 | 252 | 86 | |||
XM_005659422.3 | XP_005659479.2 | 250 | 86 | |||
XM_005659423.3 | XP_005659480.2 | 245 | 86 | |||
GenBank mRNA | AK238036.1 | - | 2 - 3 | 183 | 86 | |
AK399976.1 | - | 2 - 3 | 195 | 86 | ||
FD593970.1 | - | 2 - 3 | 256 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |