Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244521.1 | NP_001231450.1 | 4 - 5 | 798 | 64 | |
GenBank mRNA | AK237645.1 | - | 4 - 5 | 807 | 64 | |
AY336601.2 | - | 3 - 4 | 400 | 64 | ||
FD641154.1 | - | 3 - 4 | 352 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |