Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244530.1 | NP_001231459.1 | 4 - 5 | 673 | 75 | |
XM_005664650.2 | XP_005664707.1 | 647 | 75 | |||
XM_021093375.1 | XP_020949034.1 | 609 | 75 | |||
GenBank mRNA | AK240174.1 | - | 4 - 5 | 554 | 75 | |
AK240588.1 | - | 4 - 5 | 673 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |