Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244632.1 | NP_001231561.1 | 5 - 6 | 740 | 82 | |
XM_005664982.3 | XP_005665039.1 | 815 | 82 | |||
XM_005664983.3 | XP_005665040.1 | 744 | 82 | |||
XM_005664985.3 | XP_005665042.1 | 636 | 82 | |||
XM_013998960.2 | XP_013854414.1 | 910 | 82 | |||
XM_021093349.1 | XP_020949008.1 | 816 | 82 | |||
XM_021093350.1 | XP_020949009.1 | 1070 | 82 | |||
XM_021093351.1 | XP_020949010.1 | 900 | 82 | |||
XM_021093352.1 | XP_020949011.1 | 910 | 82 | |||
XM_021093353.1 | XP_020949012.1 | 648 | 82 | |||
XM_021093354.1 | XP_020949013.1 | 770 | 82 | |||
XM_021093355.1 | XP_020949014.1 | 1024 | 82 | |||
XM_021093356.1 | XP_020949015.1 | 748 | 82 | |||
XM_021093357.1 | XP_020949016.1 | 812 | 82 | |||
XM_021093358.1 | XP_020949017.1 | 1069 | 82 | |||
XM_021093359.1 | XP_020949018.1 | 1023 | 82 | |||
XM_021093360.1 | XP_020949019.1 | 2179 | 82 | |||
XM_021093361.1 | XP_020949020.1 | 999 | 82 | |||
GenBank mRNA | AK235463.1 | - | 5 - 6 | 775 | 82 | |
AK350891.1 | - | 5 - 6 | 742 | 82 | ||
AK352350.1 | - | 5 - 6 | 740 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |