Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244720.1 | NP_001231649.1 | 4 - 5 | 247 | 80 | |
GenBank mRNA | AK237926.1 | - | 3 - 4 | 386 | 80 | |
AK239005.1 | - | 3 - 4 | 355 | 80 | ||
AK239795.1 | - | 4 - 5 | 251 | 80 | ||
AK393503.1 | - | 247 | 80 | |||
AK399538.1 | - | 5 - 6 | 468 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |