Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244786.1 | NP_001231715.1 | 1 - 2 | 312 | 106 | |
GenBank mRNA | AK231845.1 | - | 1 - 2 | 314 | 106 | |
AK239323.1 | - | 1 - 2 | 319 | 106 | ||
AY610052.1 | - | 1 - 2 | 301 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |