Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB081803.1 | - | 1350 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Triticum aestivum |