Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB158409.1 | - | 747 | 76 | ||
AB161682.1 | - | 747 | 76 |