Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF334185.1 | - | 351 | 102 | ||
AY754742.1 | - | 272 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Triticum aestivum |