Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ013358.1 | - | 649 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Triticum aestivum |