Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EF397613.1 | - | 750 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Triticum aestivum |