Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ237993.1 | - | 5 - 6 | 539 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Vitis vinifera Probe spans exons |