Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ490336.1 | - | 8 - 9 | 1498 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Vitis vinifera Probe spans exons |