Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ865335.1 | - | 4 - 5 | 778 | 75 | |
DQ129685.1 | - | 4 - 5 | 778 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |