Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY337615.1 | - | 1 - 2 | 518 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Vitis vinifera Probe spans exons |