Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY538260.1 | - | 1 - 2 | 159 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Vitis vinifera Probe spans exons |