Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY538261.1 | - | 4 - 5 | 928 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |