Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP2;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ834699.1 | - | 3 - 4 | 745 | 79 | |
EF364436.1 | - | 3 - 4 | 769 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |